Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441368

Sigma-Aldrich

Deuterium oxide

"100%", 99.96 atom % D

Synonym(s):

Heavy water, Water-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
D2O
CAS Number:
Molecular Weight:
20.03
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

Quality Level

isotopic purity

99.96 atom % D

packaging

pk of 10 × 0.75 mL

bp

101.4 °C (lit.)

mp

3.8 °C (lit.)

SMILES string

[2H]O[2H]

InChI

1S/H2O/h1H2/i/hD2

InChI key

XLYOFNOQVPJJNP-ZSJDYOACSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deuterium oxide (D2O) also known as heavy water is deuterated water. It is safe and inexpensive source of deuterium (D) for the D/H exchange reactions.[1] The equilibrium constant for the exchange of deuterium between hydrogen (H) and D2O molecules has been determined.[2] Reports suggest that gelatin gels prepared in D2O show higher rigidity than gels prepared in water.[3] The influence of temperature on the heat capacity of deuterium in crystalline and liquid state was studied calorimetrically and its melting point, enthalpy and entropy of fusion were determined.[4] Its utility as a potential therapeutic agent in treating human pancreatic cancer has been studied.[5]

Application

Deuterium oxide may be used in the following processes:
  • As an NMR solvent.[6]
  • Preparation of β-deuterated alcohols.[1]
  • For the adsorption of neutrons in atomic reactors.[4]
  • Deuterium transfer agent in combination with hexamethyldisilane in the preparation of 1,2-dideuterioalkenes from alkynes.[7]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Exchange Reaction of Hydrogen and Deuterium Oxide.
Crist RH and Dalin GA
J. Chem. Phys. , 2(8), 548-548 (1934)
NMR spectroscopy of saccharide-doped PAGAT dosimeters.
Skyt PS, et al.
Journal of Physics. Conference Series, 573 (2015)
Ruthenium-Catalyzed Regioselective Deuteration of Alcohols at the β-Carbon Position with Deuterium Oxide.
Tse SKS, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 17(49), 13918-13925 (2011)
Thermodynamic properties of deuterium oxide in the temperature range from 6 to 350K.
Smirnova NN, et al.
The Journal of Chemical Thermodynamics, 38(7), 879-883 (2006)
Gelatin gels in deuterium oxide.
Oakenfull D and Scott A.
Food Hydrocolloids, 17(2), 207-210 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service