441317
Acetonitrile-d3
99.8 atom % D
Synonym(s):
Methyl-d3 cyanide, Trideuteroacetonitrile
About This Item
isotopic purity
99.8 atom % D
assay
≥99.00%
form
liquid
expl. lim.
3.0-16 % (lit.)
packaging
pk of 10 × 0.75 mL
impurities
≤0.0200% water
water
refractive index
n20/D 1.341 (lit.)
bp
80.7 °C (lit.)
mp
-46 °C (lit.)
density
0.844 g/mL at 25 °C (lit.)
mass shift
M+3
SMILES string
[2H]C([2H])([2H])C#N
InChI
1S/C2H3N/c1-2-3/h1H3/i1D3
InChI key
WEVYAHXRMPXWCK-FIBGUPNXSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
35.6 °F - closed cup
flash_point_c
2.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service