439886
Poly(1,2-butylene glycol) monobutyl ether
average Mn ~500
About This Item
mol wt
average Mn ~500
degree of polymerization ~5
viscosity
18 cSt(40 °C)(lit.)
transition temp
pour point −56 °C
solubility
oil: soluble
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service