Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43970

Sigma-Aldrich

trans-8, trans-10-Dodecadienol

≥97.0% (GC)

Synonym(s):

(E,E)-8,10-DDDOL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCH2(CH2)5CH2OH
CAS Number:
Molecular Weight:
182.30
Beilstein/REAXYS Number:
2325636
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0% (GC)

mp

30-32 °C

storage temp.

2-8°C

SMILES string

C\C=C\C=C\CCCCCCCO

InChI

1S/C12H22O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-5,13H,6-12H2,1H3/b3-2+,5-4+

InChI key

CSWBSLXBXRFNST-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service