Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439118

Sigma-Aldrich

O-(2-Chlorophenyl) dichlorothiophosphate

97%

Synonym(s):

O-(2-Chlorophenyl) phosphorodichloridothionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OP(S)Cl2
CAS Number:
Molecular Weight:
261.49
Beilstein/REAXYS Number:
1960516
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.587 (lit.)

bp

242 °C (lit.)

density

1.505 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1OP(Cl)(Cl)=S

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service