Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438324

Sigma-Aldrich

Tetramethylammonium hydrogenphthalate

≥99.99%

Synonym(s):

Phthalic acid mono(tetramethylammonium)salt, Tetramethylammonium phthalate monobasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N[O2CC6H4-2-(CO2H)]
CAS Number:
Molecular Weight:
239.27
Beilstein/REAXYS Number:
4621101
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

≥99.99%

mp

148-150 °C (lit.)

SMILES string

C[N+](C)(C)C.OC(=O)c1ccccc1C([O-])=O

InChI

1S/C8H6O4.C4H12N/c9-7(10)5-3-1-2-4-6(5)8(11)12;1-5(2,3)4/h1-4H,(H,9,10)(H,11,12);1-4H3/q;+1/p-1

InChI key

KQTFRASJMFIJPC-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service