Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437328

Sigma-Aldrich

(S)-(+)-Pantolactone

99%

Synonym(s):

(S)-(+)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (S)-(+)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (S)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, L-Pantolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
Beilstein/REAXYS Number:
4659425
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

99%

optical activity

[α]20/D +53°, c = 2.4 in H2O

bp

120-122 °C/15 mmHg (lit.)

mp

89-93 °C

SMILES string

CC1(C)COC(=O)[C@H]1O

InChI

1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m1/s1

InChI key

SERHXTVXHNVDKA-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the resolution of ketoprofen, a non-steroidal, anti-inflammatory drug.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calmes, M. et al.
Tetrahedron, 50, 6875-6875 (1994)
Optical resolution of pantolactone by a novel fungal enzyme, lactonohydrolase.
S Shimizu et al.
Annals of the New York Academy of Sciences, 799, 650-658 (1996-10-12)
A G Moĭseenok et al.
Voprosy meditsinskoi khimii, 30(1), 126-128 (1984-01-01)
A gas chromatographic procedure is described for quantitative estimation of alpha-hydroxy-beta 1 beta-dimethyl-gamma-butyrolactone (pantolactone). The chromatography was carried out using "LHM-8MD" apparatus equipped with a flame ionization detector. Columns with chromaton N-AW modified by 5% silicone XE-60 were used, helium
Bing Chen et al.
Applied biochemistry and biotechnology, 162(3), 744-756 (2009-10-31)
The levo-lactonase gene of Fusarium proliferatum ECU2002 (EC3.1.1.25) was cloned and expressed in Escherichia coli JM109 (DE3) for biocatalytic resolution of industrially important chiral lactones, including DL-pantoyl lactone which was a key precursor to calcium D-pantothenate. By increasing the biomass
Atul K Hajare et al.
The Journal of organic chemistry, 76(3), 963-966 (2011-01-05)
An efficient synthesis of potent molluscicidal agent cyanolide A, a glycosidic 16-membered macrolide, starting from D-(-)-pantolactone is reported. Highly stereoselective aldol, oxa-Michael addition, and Yamaguchi macrolactonization are the key steps in the present synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service