Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437158

Sigma-Aldrich

2,6-Dimethyl-1,3-dioxan-4-ol acetate, mixture of cis and trans

technical grade, 85%

Synonym(s):

Dimethoxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

assay

85%

refractive index

n20/D 1.433 (lit.)

bp

66-68 °C/3 mmHg (lit.)

density

1.067 g/mL at 25 °C (lit.)

SMILES string

CC1CC(OC(C)O1)OC(C)=O

InChI

1S/C8H14O4/c1-5-4-8(11-6(2)9)12-7(3)10-5/h5,7-8H,4H2,1-3H3

InChI key

PHMNXPYGVPEQSJ-UHFFFAOYSA-N

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concomitant photocontact allergy to a benzophenone derivative and a sunscreen preservative, 6-acetoxy-2,4,-dimethyl-m-dioxane.
K Kalimo et al.
Photo-dermatology, 1(6), 315-317 (1984-12-01)
E Shmunes et al.
Contact dermatitis, 6(6), 421-424 (1980-10-01)
A textile worker exposed to dimethoxane in an aqueous system developed allergic contact dermatitis. Patch testing to this compound and its possible products in an aqueous system demonstrated sensitization to acetaldehyde and crotonaldehyde.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service