Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436968

Sigma-Aldrich

Trimethylsilyl nonafluoro-1-butanesulfonate

98%

Synonym(s):

Trimethylsilyl nonaflate, Trimethylsilyl perfluoro-1-butanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9F9O3SSi
CAS Number:
Molecular Weight:
372.28
Beilstein/REAXYS Number:
2166535
MDL number:
UNSPSC Code:
12352103

assay

98%

refractive index

n20/D 1.344 (lit.)

bp

61 °C/13 mmHg (lit.)

density

1.438 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

application

Employed in the single-step preparation of mixed iodosobenzenesulfonates.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 31, 4821-4821 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service