Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436879

Sigma-Aldrich

Borane tetrahydrofuran complex solution

1.5 M in THF and ether, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BH3OC4H8
CAS Number:
Molecular Weight:
85.94
Beilstein/REAXYS Number:
3668402
MDL number:
UNSPSC Code:
12352000

grade

technical grade

concentration

1.5 M in THF and ether

bp

35 °C

density

0.846 g/mL at 25 °C

SMILES string

B.C1CCOC1

InChI

1S/C4H8O.BH3/c1-2-4-5-3-1;/h1-4H2;1H3

InChI key

RMCYTHFAWCWRFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service