About This Item
assay
97%
refractive index
n20/D 1.4530 (lit.)
bp
211-212 °C (lit.)
density
1.330 g/mL at 25 °C (lit.)
SMILES string
ClC(=O)OCCCCOC(Cl)=O
InChI
1S/C6H8Cl2O4/c7-5(9)11-3-1-2-4-12-6(8)10/h1-4H2
InChI key
AUKORMQZRYQYGI-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service