436046
Calcium fluorophosphate dihydrate
Synonym(s):
Calcium monofluorophosphate dihydrate
About This Item
form
powder
solubility
organic solvents: insoluble(lit.)
SMILES string
O.O.[Ca++].[O-]P([O-])(F)=O
InChI
1S/Ca.FH2O3P.2H2O/c;1-5(2,3)4;;/h;(H2,2,3,4);2*1H2/q+2;;;/p-2
InChI key
NBTASZQYZYVJAU-UHFFFAOYSA-L
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service