435708
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride
72%
Synonym(s):
AEM Q9-6346, DMOAP
About This Item
Recommended Products
assay
72%
impurities
13 wt. % methyl alcohol
15 wt. % (3-chloropropyl)trimethoxysilane
density
0.89 g/mL at 20 °C (lit.)
SMILES string
[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC
InChI
1S/C26H58NO3Si.ClH/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-24-27(2,3)25-23-26-31(28-4,29-5)30-6;/h7-26H2,1-6H3;1H/q+1;/p-1
InChI key
WSFMFXQNYPNYGG-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 1
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
93.0 °F - closed cup
flash_point_c
33.9 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service