Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

433284

Sigma-Aldrich

Formaldehyde solution

35 wt. % in H2O

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein/REAXYS Number:
1209228
MDL number:
Pricing and availability is not currently available.

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

autoignition temp.

572 °F

availability

available only in EU

concentration

35 wt. % in H2O

impurities

10% methyl alcohol

density

1.09 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Buchholz et al.
Antimicrobial agents and chemotherapy, 54(1), 312-319 (2009-10-14)
Chip calorimetry is introduced as a new monitoring tool that provides real-time information about the physiological state of biofilms. Its potential for use for the study of the effects of antibiotics and other biocides was tested. Established Pseudomonas putida biofilms
Denis Fourches et al.
Chemical research in toxicology, 23(1), 171-183 (2009-12-18)
Drug-induced liver injury is one of the main causes of drug attrition. The ability to predict the liver effects of drug candidates from their chemical structures is critical to help guide experimental drug discovery projects toward safer medicines. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service