Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

431664

Sigma-Aldrich

Methyl-d3 tribromoacetate

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CBr3CO2CD3
CAS Number:
Molecular Weight:
313.79
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Quality Level

refractive index

n20/D 1.558 (lit.)

bp

103 °C/15 mmHg (lit.)

density

2.353 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(=O)C(Br)(Br)Br

InChI

1S/C3H3Br3O2/c1-8-2(7)3(4,5)6/h1H3/i1D3

InChI key

QQHSHLKOWBDOFC-FIBGUPNXSA-N

Related Categories

Application

For the determination of tribromomethane in wastewater.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service