Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430129

Sigma-Aldrich

Poly(acrylonitrile-co-butadiene-co-styrene)

acrylonitrile ~25 wt. %, melt index 1-3 g/10 min (230°C/3.8kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CN)]x(CH2CH=CHCH2)y[CH2CH(C6H5)]Z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

melt index

1-3 g/10 min (230°C/3.8kg)

composition

acrylonitrile, ~25 wt. %

impact strength

7.0 ft-lb/in. (Izod, ASTM D 256, 73 °F, notched)

hardness

105 (Rockwell R, ASTM D 785)

solubility

acetone, MEK and DMF: soluble
water and alcohols: insoluble

density

1.05 g/mL at 25 °C

SMILES string

C=CC=C.C=CC#N.C=Cc1ccccc1

Looking for similar products? Visit Product Comparison Guide

Application

General purpose molding polymer.

Features and Benefits

Good impact, resiliency and good moldability.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chul-Hyun Park et al.
Environmental science & technology, 42(1), 249-255 (2008-03-21)
Plastics are widely used in everyday life as a useful material, and thus their consumption is growing at a rate of about 5% per year in Korea. However, the constant generation of plastic wastes and their disposal generates environmental problems
Chia-Yuan Chang et al.
Bioresource technology, 102(9), 5325-5330 (2010-12-31)
This study investigated the feasibility and the treatment efficiency of a cyclic anoxic/aerobic two-stage MBR for treating polymeric industrial wastewater. The anoxic/aerobic hybrid MBR was operated without sludge withdrawal except sampling during the study. The results showed that the highest
Bo Lai et al.
Journal of chromatography. A, 1244, 161-167 (2012-05-25)
Acrylonitrile-butadiene-styrene (ABS) resin manufacturing wastewater is a complicated, toxic and refractory industrial wastewater. Comprehensive and accurate analysis of the typical pollutants in ABS resin manufacturing wastewater is critical to develop cost-effective wastewater treatment technologies. In this paper, a comprehensively qualitative
Hui Wang et al.
Waste management (New York, N.Y.), 32(7), 1297-1305 (2012-04-17)
The aim of this research was to separate waste plastics acrylonitrile butadiene styrene (ABS) and polystyrene (PS) by dissolved air flotation in a self-designed dissolved air flotation apparatus. The effects of wetting agents, frother, conditioning time and flotation time on
Chun-Chin Wang et al.
Journal of hazardous materials, 145(1-2), 136-141 (2006-12-13)
epsilon-Caprolactam has high COD and toxicity, so its discharge to natural water and soil systems may lead to an adverse environmental effect on water quality, endangering public health and welfare. This investigation attempts to isolate epsilon-caprolactam denitrifying bacteria from a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service