Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429805

Sigma-Aldrich

Methyl (S)-(−)-2-isocyanato-3-methylbutyrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NCO)CO2CH3
CAS Number:
Molecular Weight:
157.17
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

99%

optical activity

[α]20/D −22°, neat

refractive index

n20/D 1.428 (lit.)

bp

75 °C/0.04 mmHg (lit.)

density

1.053 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@@H](N=C=O)C(C)C

InChI

1S/C7H11NO3/c1-5(2)6(8-4-9)7(10)11-3/h5-6H,1-3H3/t6-/m0/s1

InChI key

JCVZXXSBCISMLJ-LURJTMIESA-N

Application

Precursor for the synthesis of molecular scaffolds[1] and bridged azapeptide mimetics.[2]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

172.4 °F - closed cup

flash_point_c

78 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norwick, J.S. et al.
Journal of the American Chemical Society, 117, 89-89 (1995)
Gante, J. et al.
The Journal of Organic Chemistry, 59, 6487-6487 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service