Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429139

Sigma-Aldrich

Poly(ethylene-co-propylene)

melt index 30 g/10 min (230°C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH3)]X(CH2CH2)Y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

pellets

melt index

30 g/10 min (230°C/2.16kg)

composition

1-3 wt. % (ethylene)

hardness

80 (Shore D, ASTM D 2240)

transition temp

Tm 128 °C

density

0.9 g/mL at 25 °C (lit.)

SMILES string

C=C.CC=C

InChI

1S/C3H6.C2H4/c1-3-2;1-2/h3H,1H2,2H3;1-2H2

InChI key

HQQADJVZYDDRJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Injection molded thin-walled parts.

Features and Benefits

Random copolymer. Low warpage and good film clarity, toughness and resistance to chemical and detergent attack.

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service