Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428388

Sigma-Aldrich

Diglycidyl 1,2,3,6-tetrahydrophthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O6
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
12162002

vapor pressure

<10-4 ( 20 °C)

eq. wt.

154-169 Epoxide

viscosity

450 cP(25 °C)(lit.)

SMILES string

O=C(OCC1CO1)C2CC=CCC2C(=O)OCC3CO3

application

Vacuum-pressure or full bath impregnation for motors and generators with medium to high power ratings.

Features and Benefits

Polymerized monomer has high impact and shear strength, arc and tracking resistance and good thermal aging resistance with glass cloth and mica paper.

Physical form

Difunctional epoxy monomer.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service