Skip to Content
MilliporeSigma
All Photos(1)

Documents

42823

Sigma-Aldrich

2,3-Diphenyl-1,3-butadiene

technical, ~90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(C6H5)C(C6H5)=CH2
CAS Number:
Molecular Weight:
206.28
Beilstein/REAXYS Number:
1363613
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

~90% (HPLC)

mp

40-45 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

C=C(c1ccccc1)C(=C)c2ccccc2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service