Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423572

Sigma-Aldrich

Boc-Ala-OMe

99%

Synonym(s):

N-(tert-Butoxycarbonyl)-L-alanine methyl ester, Boc-L-alanine methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
203.24
Beilstein/REAXYS Number:
1872545
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

liquid

optical activity

[α]21/D −45°, c = 1 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

32-35 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](C)NC(=O)OC(C)(C)C

InChI

1S/C9H17NO4/c1-6(7(11)13-5)10-8(12)14-9(2,3)4/h6H,1-5H3,(H,10,12)/t6-/m0/s1

InChI key

GJDICGOCZGRDFM-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service