Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423408

(S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

99%, ChiraSelect

Synonym(s):

(S)-(+)-MTPA-Cl, Mosher’s acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Beilstein/REAXYS Number:
3591562
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

product line

ChiraSelect

assay

99%

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.469 (lit.)

bp

213-214 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CO[C@@](C(Cl)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H8ClF3O2/c1-16-9(8(11)15,10(12,13)14)7-5-3-2-4-6-7/h2-6H,1H3/t9-/m1/s1

InChI key

PAORVUMOXXAMPL-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Miller et al.
Journal of chromatography, 307(2), 335-342 (1984-05-11)
High-performance liquid chromatography (HPLC) was employed for resolution of enantiomers of chiral ring-substituted 1-phenyl-2- aminopropanes (amphetamines) and 1-phenylethylamine following derivatization with four chiral reagents: (R)-(+)-1-phenylethyl isocyanate ( PEIC ), (-)-alpha-methoxy-alpha-(trifluoromethyl)phenylacetyl chloride ( MTPA X Cl), 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate ( GITC )
Yanhong Ma et al.
Marine drugs, 13(10), 6306-6318 (2015-10-17)
Six new compounds with polyketide decalin ring, peaurantiogriseols A-F (1-6), along with two known compounds, aspermytin A (7), 1-propanone,3-hydroxy-1- (1,2,4a,5,6,7,8,8a-octahydro-2,5-dihydroxy-1,2,6-trimethyl-1-naphthalenyl) (8), were isolated from the fermentation products of mangrove endophytic fungus Penicillium aurantiogriseum 328#. Their structures were elucidated based on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service