Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423106

Sigma-Aldrich

Acetonitrile-d3

"100%", 99.96 atom % D

Synonym(s):

Methyl-d3 cyanide, Trideuteroacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CN
CAS Number:
Molecular Weight:
44.07
Beilstein/REAXYS Number:
1740230
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.96 atom % D

assay

99% (CP)

packaging

pk of 10 × 1 mL

refractive index

n20/D 1.341 (lit.)

bp

80.7 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C#N

InChI

1S/C2H3N/c1-2-3/h1H3/i1D3

InChI key

WEVYAHXRMPXWCK-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro G Lichtscheidl et al.
Dalton transactions (Cambridge, England : 2003), 44(36), 16156-16163 (2015-08-22)
The synthesis and full characterization, including Nuclear Magnetic Resonance (NMR) data ((1)H, (13)C{(1)H} and (119)Sn{(1)H}), for a series of Me3SnX (X = O-2,6-(t)Bu2C6H3 (), (Me3Sn)N(2,6-(i)Pr2C6H3) (), NH-2,4,6-(t)Bu3C6H2 (), N(SiMe3)2 (), NEt2, C5Me5 (), Cl, Br, I, and SnMe3) compounds in
Anne Warner et al.
Journal of pharmaceutical and biomedical analysis, 105, 46-54 (2014-12-22)
Stability-indicating reversed phase HPLC methods have been developed and validated for the determination of 13 potential process and degradation impurities in pemetrexed disodium drug substance (DS) and pemetrexed for injection drug product (DP). This paper describes the development of HPLC-UV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service