Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421820

Sigma-Aldrich

(1S)-(−)-Camphanic chloride

≥98%, ChiraSelect

Synonym(s):

(−)-Camphanoyl chloride, (1S)-3-Oxo-4,7,7-trimethyl-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H13O3Cl
CAS Number:
Molecular Weight:
216.66
Beilstein/REAXYS Number:
3590860
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

product line

ChiraSelect

assay

≥98%

optical purity

ee: 99% (GLC)

mp

71-73 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(Cl)=O

InChI

1S/C10H13ClO3/c1-8(2)9(3)4-5-10(8,6(11)12)14-7(9)13/h4-5H2,1-3H3/t9-,10+/m0/s1

InChI key

PAXWODJTHKJQDZ-VHSXEESVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Resolving agent for racemic alcohols.

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 2899-2899 (1992)
The determination of the absolute configurations of diastereomers of (S)-camphanoyl 3-hydroxy-5-oxohexanoic acid derivatives by X-ray crystallography.
Kwai-Ming Cheung et al.
Angewandte Chemie (International ed. in English), 41(7), 1198-1202 (2002-12-20)
M Ruud-Christensen et al.
Journal of chromatography, 491(2), 355-366 (1989-07-21)
A reversed-phase high-performance liquid chromatographic assay has been developed for determination of (R)-(--)-and (S)-(+)-proxyphylline in human plasma. The procedure is based on liquid-solid extraction of proxyphylline from plasma followed by derivatization of extracted proxyphylline with (--)-camphanoyl chloride. The ratio between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service