Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421782

Sigma-Aldrich

(S)-(+)-1-(1-Naphthyl)ethyl isocyanate

≥99%, ChiraSelect

Synonym(s):

(S)-NEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NCO
CAS Number:
Molecular Weight:
197.23
Beilstein/REAXYS Number:
4675071
MDL number:

product line

ChiraSelect

assay

≥99%

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.604 (lit.)

bp

106-108 °C/0.16 mmHg (lit.)

density

1.128 g/mL at 25 °C (lit.)

SMILES string

C[C@H](N=C=O)c1cccc2ccccc12

InChI

1S/C13H11NO/c1-10(14-9-15)12-8-4-6-11-5-2-3-7-13(11)12/h2-8,10H,1H3/t10-/m0/s1

Inchi Key

GONOHGQPZFXJOJ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service