Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420956

Sigma-Aldrich

QUILON® L chromium complex solution

8.5-9.5 wt. % Cr in isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

concentration

8.5-9.5 wt. % Cr in isopropanol

bp

82 °C

mp

4 °C

density

1.025 g/mL at 25 °C

SMILES string

[Cr]

Legal Information

DuPont Product
QUILON is a registered trademark of E. I. du Pont de Nemours and Company

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

50.0 °F - closed cup

flash_point_c

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service