Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

419893

Sigma-Aldrich

Trimethylsilyl propiolate

97%

Synonym(s):

Trimethylsilyl acetylenecarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CCO2Si(CH3)3
CAS Number:
Molecular Weight:
142.23
Beilstein/REAXYS Number:
2237544
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.416 (lit.)

bp

70-75 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OC(=O)C#C

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G Micetich et al.
Journal of medicinal chemistry, 30(8), 1469-1474 (1987-08-01)
Benzhydryl 2 beta-[(1,2,3-triazol-1-yl)methyl]-2 alpha-methylpenam- 3 alpha-carboxylate 1,1-dioxide was prepared by heating benzhydryl 2 beta-(azidomethyl)-2 alpha-methylpenam-3 alpha-carboxylate 1,1-dioxide with (trimethylsilyl)acetylene. The ester group was removed by hydrogenolysis to give sodium 2 beta-[(1,2,3-triazol-1-yl)methyl]-2 alpha-methylpenam-3 alpha-carboxylate 1,1-dioxide (3i, YTR-830), which was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service