418528
Poly(1-vinylpyrrolidone-co-acrylic acid)
average Mw ~96,000 by GPC, average Mn ~20,000 by GPC, powder (Fine)
About This Item
form
powder (Fine)
mol wt
average Mn ~20,000 by GPC
average Mw ~96,000 by GPC
hardness
6 H (ASTM D 3363)
impurities
≤3.0% moisture content
color
white
relative viscosity
1.4-1.7, 1 wt. % in 0.25 M NaOH in 0.2 N LiNO3(lit.)
acid number
170‑210
transition temp
Tg 185 °C
solubility
H2O: soluble (pH ≥4.5)
Application
Features and Benefits
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service