Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417785

Sigma-Aldrich

4,4-Dimethyloxazolidine solution

75 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

concentration

75 wt. % in H2O

refractive index

n20/D 1.432

density

0.942 g/mL at 25 °C

SMILES string

CC1(C)COCN1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Zhang et al.
Biochimica et biophysica acta, 861(1), 201-204 (1986-09-25)
The effects of the phytotoxin, syringomycin, produced by Pseudomonas syringae pv. syringae, were examined on cells of a wild-type and a respiratory-deficient (rho0) mutant of Saccharomyces cerevisiae. The growth of both strains in liquid culture was inhibited by 0.5 micrograms
Mutagenicity of two non-formaldehyde-forming antimicrobial agents.
W C Monte et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 21(5), 695-696 (1983-10-01)
Youhei Fukui et al.
Antonie van Leeuwenhoek, 107(5), 1155-1163 (2015-02-25)
Three Gram-negative, aerobic, halophilic bacterial strains, SCM2-10(T), SCM-4, and 14C-6, were isolated from the algal medium of the red alga Pyropia yezoensis (previously classified as Porphyra yezoensis) grown in laboratory experiments. Phylogenetic analysis based on 16S rRNA gene sequences revealed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service