416606
Bisphenol A ethoxylate sulfate ammonium salt solution
60 wt. % in H2O, EO/phenol 15
About This Item
composition
EO/phenol, 15
concentration
60 wt. % in H2O
refractive index
n20/D 1.431
density
1.115 g/mL at 25 °C
SMILES string
OCCO.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service