416312
Poly(methyl vinyl ether-alt-maleic anhydride)
average Mw ~2,400,000 by LS, average Mn ~1,130,000
Synonym(s):
maleated copolymer, maleated polymer, maleic copolymer
About This Item
Recommended Products
mol wt
average Mn ~1,130,000
average Mw ~2,400,000 by LS
impurities
<2% benzene
solubility
H2O: soluble (upon hydrolysis)
organic solvents: soluble
InChI
1S/C4H2O3.C3H6O/c5-3-1-2-4(6)7-3;1-3-4-2/h1-2H;3H,1H2,2H3
InChI key
UPBDXRPQPOWRKR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service