416150
Pentaerythritol ethoxylate (3/4 EO/OH)
average Mn ~270
Synonym(s):
Pentaerythritol ethoxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
viscous liquid
mol wt
average Mn ~270
refractive index
n20/D 1.481
bp
>300 °C (lit.)
density
1.2 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service