Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416061

Sigma-Aldrich

Poly(acrylic acid-co-maleic acid) sodium salt

average Mw ~50,000

Synonym(s):

Acrylic acid-maleic anhydride copolymer sodium salt, Maleic anhydride-acrylic acid copolymer sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mw ~50,000

SMILES string

OC(=O)C=C.OC(=O)\C=C\C(=O)O[Na]

InChI

1S/C4H2O3.C3H4O2.Na/c5-3-1-2-4(6)7-3;1-2-3(4)5;/h1-2H;2H,1H2,(H,4,5);/q;;+1/p-1

InChI key

LDLISKWWEPKHAT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1:1 mole ratio of AA:MA

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service