About This Item
assay
95%
refractive index
n20/D 1.438 (lit.)
bp
78-80 °C (lit.)
density
0.809 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)C(=N)C(C)(C)C
InChI
1S/C9H19N/c1-8(2,3)7(10)9(4,5)6/h10H,1-6H3
InChI key
VCMKYJMEMATQPE-UHFFFAOYSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
84.2 °F - closed cup
flash_point_c
29 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service