Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414875

Sigma-Aldrich

2,4-Dichlorophenyl isothiocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCS
CAS Number:
Molecular Weight:
204.08
Beilstein/REAXYS Number:
777072
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

bp

260 °C (lit.)

mp

36-40 °C (lit.)

density

1.41 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1ccc(N=C=S)c(Cl)c1

InChI

1S/C7H3Cl2NS/c8-5-1-2-7(10-4-11)6(9)3-5/h1-3H

InChI key

WVBNZZHGECFCSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service