Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414832

Sigma-Aldrich

5-Nitro-3-pyrazolecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N3O4
CAS Number:
Molecular Weight:
157.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

188-190 °C (lit.)

SMILES string

OC(=O)c1cc([nH]n1)[N+]([O-])=O

InChI key

HKYHBMLIEAMWRO-UHFFFAOYSA-N

Caution

Material is currently undergoing additional testing for DOT shipping approval. This material will be on back order for an indefinite period of time until testing is complete.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

4.1A - Other explosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 12, 1815-1826 (2003)
Darren Dressen et al.
Journal of medicinal chemistry, 50(21), 5161-5167 (2007-09-21)
The B1 receptor is an attractive target for the treatment of pain and inflammation. A series of 3-carboxamido-5-phenacylamino pyrazole B1 receptor antagonists are described that exhibit good potency against B1 and high selectivity over B2. Initially, N-unsubstituted pyrazoles were studied
Katerina Cernovská et al.
Organic & biomolecular chemistry, 2(11), 1603-1611 (2004-05-27)
Pyrazole amino acid oligoamides were prepared on polyethylene glycol starting from nitro pyrazole carboxylic acids or protected pyrazole amino acids. The polymer support facilitates product isolation during synthesis and makes the target oligoamides soluble in chloroform and water. This allows
Discovery of novel transcription factor inhibitors using a pyrazole-based small molecule library.
Ha H and Kim BM.
Bull. Korean Chem. Soc., 29(2), 323-323 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service