Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414697

Millipore

Chloroform

capillary GC grade, ≥99.9%, contains amylenes as stabilizer

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein/REAXYS Number:
1731042
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

capillary GC grade

vapor density

4.1 (vs air)

vapor pressure

160 mmHg ( 20 °C)

assay

≥99.9%

purified by

glass distillation

contains

amylenes as stabilizer

impurities

<1 ng/mL organic residue (GC/FID, as toluene)

evapn. residue

<0.0001%

halogenated residue (GC/ECD)

<2 ng/L (as lindane)

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)
61 °C

mp

−63 °C (lit.)

density

1.48 g/mL at 25 °C
1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

Inchi Key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for environmental analysis

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sue M Ennaceur et al.
Biophysical journal, 96(4), 1399-1407 (2009-02-17)
The adsorption and insertion kinetics for the association of two 34-residue cyclic peptides with phosphocholine membranes have been studied using circular and linear dichroism approaches. The two peptides studied are identical with the exception of two residues, which are both
Shashank R Sirsi et al.
Journal of nanobiotechnology, 7, 1-1 (2009-04-09)
Antisense oligonucleotides (AOs) have been shown to induce dystrophin expression in muscles cells of patients with Duchenne Muscular Dystrophy (DMD) and in the mdx mouse, the murine model of DMD. However, ineffective delivery of AOs limits their therapeutic potential. Copolymers
Patricia Fabian et al.
PloS one, 3(7), e2691-e2691 (2008-07-17)
Recent studies suggest that humans exhale fine particles during tidal breathing but little is known of their composition, particularly during infection. We conducted a study of influenza infected patients to characterize influenza virus and particle concentrations in their exhaled breath.
Elizabeth K Elliott et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(19), 5871-5879 (2008-05-17)
The amphiphilic heptapeptides-referred to as synthetic anion transporters (SATs)-mediate chloride transport in planar lipid bilayer membranes, synthetic liposomes, and mammalian cells. The SATs described have the general formula R1(2)NCOCH2OCH2CO-(Gly)3-Pro-(Gly)3-OR2. Substitution at R1 and R2 with various aliphatic or aromatic groups
Michal Legiewicz et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(38), 14365-14370 (2008-09-09)
Nuclear export of certain HIV-1 mRNAs requires an interaction between the viral Rev protein and the Rev response element (RRE), a structured element located in the Env region of its RNA genome. This interaction is an attractive target for both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service