Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414344

Sigma-Aldrich

B-Iodo-9-BBN

>95%

Synonym(s):

9-Iodo-9-borabicyclo[3.3.1]nonane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14BI
CAS Number:
Molecular Weight:
247.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

assay

>95%

bp

55 °C/0.2 mmHg (lit.)

density

1.568 g/mL at 25 °C (lit.)

SMILES string

IB1[C@@H]2CCC[C@H]1CCC2

comparable product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service