41390
(1R,exo,exo)-3-[N-(3,5-Dimethylphenyl)benzenesulfonamido]isoborneol
≥98.5% (sum of enantiomers, HPLC)
Synonym(s):
(1R,2S,3R)-(+)-3-[N-Benzenesulfonyl-N-(3,5-dimethylphenyl)amino]-2-bornanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
assay
≥98.5% (sum of enantiomers, HPLC)
optical activity
[α]20/D +65±2°, c = 4% in chloroform
mp
122-125 °C (lit.)
122-125 °C
SMILES string
Cc1cc(C)cc(c1)N([C@H]2[C@@H](O)[C@]3(C)CC[C@H]2C3(C)C)S(=O)(=O)c4ccccc4
Looking for similar products? Visit Product Comparison Guide
Other Notes
Auxiliary for very efficient asymmetric reactions: asymmetric alkylation of its esters; asymmetric conjugate addition to its enoates; hydroxylation of its esters
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Angewandte Chemie (International Edition in English), 96, 59-59 (1984)
Tetrahedron Letters, 26, 6051-6051 (1985)
Tetrahedron Letters, 26, 6047-6047 (1985)
Helvetica Chimica Acta, 69, 615-615 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service