Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412341

Sigma-Aldrich

PAMAM Dendrimer, ethylenediamine core, Generation -0.5 solution

20 wt. % in H2O

Synonym(s):

Ethylenediaminetetrapropionic acid tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2Na4O8
CAS Number:
Molecular Weight:
436.28
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

concentration

20 wt. % in H2O

refractive index

n20/D 1.37

bp

100 °C

SMILES string

[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CCN(CCN(CCC([O-])=O)CCC([O-])=O)CCC([O-])=O

InChI

1S/C14H24N2O8.4Na/c17-11(18)1-5-15(6-2-12(19)20)9-10-16(7-3-13(21)22)8-4-14(23)24;;;;/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24);;;;/q;4*+1/p-4

InChI key

CJVPOGPEEJPBFT-UHFFFAOYSA-J

Physical form

Contains 4 surface carboxylate groups

Other Notes

NOT to be used for testing in vivo in either humans or animals

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service