Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412112

Sigma-Aldrich

Bisphenol A ethoxylate dimethacrylate

average Mn ~540, EO/phenol 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2(CH2CH2O)nC6H4-4-]2C(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~540

contains

500 ppm monomethyl ether hydroquinone as inhibitor

composition

EO/phenol, 2

refractive index

n20/D 1.532

density

1.12 g/mL at 25 °C (lit.)

SMILES string

OCCO.OC(=O)C=C.CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.C3H4O2.C2H6O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;1-2-3(4)5;3-1-2-4/h3-10,16-17H,1-2H3;2H,1H2,(H,4,5);3-4H,1-2H2

Inchi Key

HGARWRTUWINPJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine H Dickens et al.
European journal of oral sciences, 112(5), 452-457 (2004-10-02)
An experimental resin-based bioactive calcium phosphate cement, intended as a pulp capping and basing material, was evaluated for dentin shear bond strength and microleakage. The interfacial morphology was examined by scanning electron microscopy (SEM). For microleakage, dentin cavities without (Group
Catherine Fan et al.
Dental materials : official publication of the Academy of Dental Materials, 27(4), 322-328 (2010-11-30)
To demonstrate that silver nanoparticles (AgNPs) could be synthesized in situ in acrylic dental resins. Light-cure (LC; bisphenol A glycidyl methacrylate, tetraethyleneglycol dimethacrylate, bisphenol A ethoxylate dimethacrylate blend) and chemical-cure systems (CC; orthodontic denture resin) were used to synthesize AgNPs
Salvatore Sauro et al.
Dental materials : official publication of the Academy of Dental Materials, 27(6), 563-572 (2011-04-02)
To compare microtensile bond strengths (MTBS) subsequent to load cycling of resin bonded acid-etched or EDTA-treated dentin using a modified ethanol wet-bonding technique. Flat dentin surfaces were obtained from extracted human molars and conditioned using 37% H(3)PO(4) (PA) (15s) or
S E Langhorst et al.
Dental materials : official publication of the Academy of Dental Materials, 25(7), 884-891 (2009-02-14)
This study explores the efficacy of an experimental orthodontic amorphous calcium phosphate (ACP) composite to remineralize in vitro subsurface enamel lesions microradiographically similar to those seen in early caries. Lesions were artificially created in extracted human molars. Single tooth sections
Milena Cadenaro et al.
Dental materials : official publication of the Academy of Dental Materials, 24(9), 1194-1200 (2008-03-18)
To evaluate the degree of conversion of five experimental adhesive systems in relation to their hydrophilicity. The resin blends ranged from hydrophobic to hydrophilic and were tested as neat bonding agents, or solvated with increasing percentages of ethanol. The hypothesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service