Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

411175

Sigma-Aldrich

Phenyl trans-styryl sulfone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHSO2C6H5
CAS Number:
Molecular Weight:
244.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

mp

74-75 °C (lit.)

SMILES string

O=S(=O)(\C=C\c1ccccc1)c2ccccc2

General description

Phenyl trans-styryl sulfone is vinyl sulfone drug.[1]

Application

Phenyl trans-styryl sulfone was employed as vinyl sulfone drug in the selective kidney disease medium (SKDM-2) for the bacterial growth.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ponnerassery S Sudheesh et al.
Diseases of aquatic organisms, 78(2), 115-127 (2008-02-22)
Renibacterium salmoninarum, the causative agent of bacterial kidney disease in salmonid fishes, is a Gram-positive diplococcobacillus belonging to the family Micrococcaceae. Analysis of the genome sequence of the bacterium demonstrated the presence of a sortase homolog (srtD), a gene specifying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service