Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

410683

Sigma-Aldrich

tert-Butyl(dichloromethyl)dimethylsilane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2CHCl2
CAS Number:
Molecular Weight:
199.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

98%

bp

70 °C/20 mmHg (lit.)

mp

42-45 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)[Si](C)(C)C(Cl)Cl

InChI

1S/C7H16Cl2Si/c1-7(2,3)10(4,5)6(8)9/h6H,1-5H3

InChI key

IIGQYABNVUNASW-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service