Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

410144

Sigma-Aldrich

Praseodymium(III) isopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr(OCH(CH3)2)3
CAS Number:
Molecular Weight:
318.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

SMILES string

CCCO[Pr](OCCC)OCCC

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service