Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407143

Sigma-Aldrich

N-(2-Furoyl)glycine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO4
CAS Number:
Molecular Weight:
169.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

98%

mp

163-165 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CNC(=O)c1ccco1

InChI

1S/C7H7NO4/c9-6(10)4-8-7(11)5-2-1-3-12-5/h1-3H,4H2,(H,8,11)(H,9,10)

InChI key

KSPQDMRTZZYQLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Parkash et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 32(10), 887-895 (1994-10-01)
The fate of furfural (2-furancarboxaldehyde) was investigated in male and female Fischer 344 (F344) rats given single oral doses of 1, 10 and 60 mg/kg and male and female CD1 mice given 1, 20 and 200 mg/kg [carbonyl-14C]furfural. There was
Deborah B Diercks et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 19(1), 18-23 (2012-01-10)
Worsening renal function in patients admitted with heart failure is associated with increased morbidity. These changes are not usually apparent initially and often take up to 48 hours to be detected. Using the novel technique of metabolomic analysis, this study
Daniel McGill et al.
Faraday discussions, 218(0), 395-416 (2019-05-23)
Metabolite identification and annotation procedures are necessary for the discovery of biomarkers indicative of phenotypes or disease states, but these processes can be bottlenecked by the sheer complexity of biofluids containing thousands of different compounds. Here we describe low-cost novel
The identification and metabolic origin of 2-furoylglycine and 2,5-furandicarboxylic acid in human urine.
J E Pettersen et al.
Clinica chimica acta; international journal of clinical chemistry, 41, 199-207 (1972-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service