Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

403385

Sigma-Aldrich

Titanium-copper alloy

powder, 6-12 μm

Synonym(s):

CuTi4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuTi4
CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

composition

Cu, 25%
Ti, 75%

particle size

6-12 μm

SMILES string

[Ti].[Cu]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service