Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

403199

Sigma-Aldrich

Iodotris(trifluoromethyl)germanium

97%

Synonym(s):

Tris(trifluoromethyl)germanium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)3GeI
CAS Number:
Molecular Weight:
406.56
Beilstein/REAXYS Number:
4133121
MDL number:
UNSPSC Code:
12352103

vapor pressure

115 mmHg ( 20 °C)

assay

97%

contains

copper as stabilizer

refractive index

n20/D 1.351 (lit.)

bp

72 °C (lit.)

mp

−40 °C (lit.)

density

2.074 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)[Ge](I)(C(F)(F)F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service