Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

399906

Sigma-Aldrich

Antimony(V) fluoride on graphite

extent of labeling: 50 wt. % loading

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbF5
CAS Number:
Molecular Weight:
216.75
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

powder

extent of labeling

50 wt. % loading

SMILES string

F[Sb](F)(F)(F)F

InChI

1S/5FH.Sb/h5*1H;/q;;;;;+5/p-5

InChI key

VBVBHWZYQGJZLR-UHFFFAOYSA-I

comparable product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service