Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

399590

Sigma-Aldrich

Calcium molybdate

99%, −325 mesh

Synonym(s):

Powellite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaMoO4
CAS Number:
Molecular Weight:
200.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Product 399590 is not currently sold in your country. Contact Technical Service

assay

99%

form

powder

particle size

−325 mesh

solubility

concentrated acid: slightly soluble(lit.)
ethanol: insoluble(lit.)

density

4.35 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-][Mo]([O-])(=O)=O

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service