Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

393673

Sigma-Aldrich

1-Methyl-2-aminoterephthalate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO2C)C6H3(NH2)CO2CH3
CAS Number:
Molecular Weight:
195.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

217-220 °C (lit.)

solubility

concentrated acid: soluble (HCl)(lit.)

functional group

carboxylic acid
ester

SMILES string

COC(=O)c1ccc(cc1N)C(O)=O

InChI

1S/C9H9NO4/c1-14-9(13)6-3-2-5(8(11)12)4-7(6)10/h2-4H,10H2,1H3,(H,11,12)

InChI key

QKOKLMFCKLEFDV-UHFFFAOYSA-N

General description

1-Methyl-2-aminoterephthalate (3-Amino-4-carbomethoxybenzoic acid, 3-Amino-4-methoxycarbonylbenzoic acid) is the 1-methyl ester of 2-aminoterephthalic acid.

Application

1-Methyl-2-aminoterephthalate is suitable reagent used in the synthesis of 1-methyl-2-iodoterephthalate.[1]
1-Methyl-2-aminoterephthalate may be used in the following syntheses:
  • triarylphosphine[2]
  • methyl 4-(hydroxymethyl-2-iodobenzoate[3]
  • 1-methyl-2-diphenylphosphinoterepthalate

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Re-examination of Dynamics of Polyeletrolytes in Salt-Free Dilute Solutions by Designing and Using a Novel Neutral- Charged- Neutral Reversible Polymer.
Zhou K, et al.
Macromolecules, 42(18), 7146-7154 (2009)
Cell surface engineering by a modified Staudinger reaction.
Saxon E and Bertozzi CR.
Science, 287(5460), 2007-2010 (2000)
Ganesan Vaidyanathan et al.
Nuclear medicine and biology, 38(1), 77-92 (2011-01-12)
Drug resistance to alkylator chemotherapy has been primarily attributed to the DNA repair protein alkylguanine-DNA alkyltransferase (AGT); thus, personalizing chemotherapy could be facilitated if tumor AGT content could be quantified prior to administering chemotherapy. We have been investigating the use

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service